missing translation for 'onlineSavingsMsg'
Learn More

CCL4L1/LAG-1 Antibody (RM0181-5G13), DyLight 350, Novus Biologicals™

Product Code. 18400196 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18400196 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18400196 Supplier Novus Biologicals Supplier No. NBP212119UV

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rat Monoclonal Antibody

CCL4L1/LAG-1 Monoclonal antibody specifically detects CCL4L1/LAG-1 in Mouse samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCL4L1/LAG-1
Applications ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone RM0181-5G13
Conjugate DyLight 350
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias AT744.2, C-C motif chemokine 4-like, CCL4Lsmall inducible cytokine A4-like, chemokine (C-C motif) ligand 4-like 1, LAG1CCL4L2, LAG-1chemokine (C-C motif) ligand 4-like, Lymphocyte activation gene 1 protein, Macrophage inflammatory protein 1-beta, macrophage inflammatory protein-1b2, MIP-1-beta, Monocyte adherence-induced protein 5-alpha, SCYA4L, SCYA4L1, SCYA4L2, Small-inducible cytokine A4-like
Host Species Rat
Immunogen Mouse recombinant CCL4L1/LAG-1
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins
Primary or Secondary Primary
Gene ID (Entrez) 9560
Target Species Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.