missing translation for 'onlineSavingsMsg'
Learn More

CCL4L1/LAG-1 Antibody (RM0181-5G13), Alexa Fluor™ 700, Novus Biologicals™

Product Code. 18414907 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18414907 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18414907 Supplier Novus Biologicals Supplier No. NBP212119AF700

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rat Monoclonal Antibody

CCL4L1/LAG-1 Monoclonal antibody specifically detects CCL4L1/LAG-1 in Mouse samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen CCL4L1/LAG-1
Applications ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone RM0181-5G13
Conjugate Alexa Fluor 700
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias AT744.2, C-C motif chemokine 4-like, CCL4Lsmall inducible cytokine A4-like, chemokine (C-C motif) ligand 4-like 1, LAG1CCL4L2, LAG-1chemokine (C-C motif) ligand 4-like, Lymphocyte activation gene 1 protein, Macrophage inflammatory protein 1-beta, macrophage inflammatory protein-1b2, MIP-1-beta, Monocyte adherence-induced protein 5-alpha, SCYA4L, SCYA4L1, SCYA4L2, Small-inducible cytokine A4-like
Host Species Rat
Immunogen Mouse recombinant CCL4L1/LAG-1
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins
Primary or Secondary Primary
Gene ID (Entrez) 9560
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.