missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CCL4L1/CCL4L2 Monoclonal Antibody (4G8)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA523300
This item is not returnable.
View return policy
Description
Peptide Sequence: SDPPTACCFS YTARKLPRNF VVDYYETSSL CSQPAVVFQT KRGKQVCADP SESWVQEYVY DLELN*.
This gene is one of several cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. This protein is similar to CCL4 which inhibits HIV entry by binding to the cellular receptor CCR5. The copy number of this gene varies among individuals; most individuals have 1-5 copies in the diploid genome, although rare individuals do not contain this gene at all. The human genome reference assembly contains two copies of this gene. This record represents the more telomeric gene.
Specifications
| CCL4L1/CCL4L2 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q8NHW4 | |
| CCL4L1 | |
| CCL4L1 (NP_996890, 28 a.a. ~ 93 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot | |
| 4G8 | |
| Unconjugated | |
| CCL4L1 | |
| ACT-2; AT744.2; CC chemokine ligand 4L1; C-C motif chemokine 4; C-C motif chemokine 4-like; C-C motif chemokine ligand 4 like 1; CCL4; CCL4L; CCL4L1; CCL4L2; chemokine (C-C motif) ligand 4-like 1; chemokine (C-C motif) ligand 4-like 1, telomeric; G-26 T-lymphocyte-secreted protein; HC21; LAG1; LAG-1; lymphocyte activation gene 1; Lymphocyte activation gene 1 protein; Macrophage inflammatory protein 1-beta; macrophage inflammatory protein-1b2; MIP1B; MIP-1-beta; MIP-1-beta(1-69); MIP-1-beta(3-69); Monocyte adherence-induced protein 5-alpha; PAT 744; Protein H400; SCYA4; SCYA4L; SCYA4L1; SCYA4L2; SIS-gamma; small inducible cytokine A4-like; Small-inducible cytokine A4; Small-inducible cytokine A4-like; T-cell activation protein 2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 388372 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction