missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCL4/MIP-1 beta Antibody (24006) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB271JF525
This item is not returnable.
View return policy
Description
CCL4/MIP-1 beta Monoclonal antibody specifically detects CCL4/MIP-1 beta in Human samples. It is validated for Western Blot,ELISA Standard (Matched Pair),Neutralization,ELISA Capture (Matched Antibody Pair),ELISA Detection (Matched Antibody Pair),Flow Cytometry (Intracellular Staining),CyTOF
Specifications
| CCL4/MIP-1 beta | |
| Monoclonal | |
| Janelia Fluor 525 | |
| ACT2, ACT-2, AT744.1, C-C motif chemokine 4, chemokine (C-C motif) ligand 4, G-26, G-26 T-lymphocyte-secreted protein, HC21, LAG1, LAG-1, Lymphocyte activation gene 1 protein, Macrophage inflammatory protein 1-beta, MIP1B, MIP1B1, MIP-1-beta, MIP-1-beta(1-69), PAT 744, Protein H400, SCYA2, SCYA4, secreted protein G-26, SIS-gamma, small inducible cytokine A4 (homologous to mouse Mip-1b), Small-inducible cytokine A4, T-cell activation protein 2 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human CCL4/MIP-1β, Ala24-Asn92, Accession # P13236 | |
| 0.1 mL | |
| Biologically Active Proteins, Chemokines and Cytokines | |
| 6351 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Western Blot, ELISA Standard (Matched Pair), Neutralization, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), Flow Cytometry (Intracellular Staining), CyTOF | |
| 24006 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction