Learn More
Invitrogen™ CCL3 (MIP-1 alpha) Human Uncoated ELISA Kit
Description
The Human CCL3 (MIP-1 alpha) Uncoated ELISA Kit contains pre-matched antibody pairs, and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of Human IL-5. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.
Specifications
Specifications
| Accession Number | P10147 |
| Assay Range | 31.25-2000 pg/mL |
| Assay Sensitivity | 31.25 pg/mL |
| Content And Storage | Capture Antibody: Pre-titrated, purified antibody Detection Antibody: Pre-titrated, biotin-conjugated antibody Standard: Recombinant protein for generating standard curve and calibrating samples 10X Coating Buffer: Buffer for plating the Capture Antibody 5X ELISA/ELISPOT Diluent: Buffer for blocking and diluting the Detection Antibody and Enzyme Enzyme: Pre-titrated Avidin-HRP Substrate: 1X TMB Solution Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards 96 Well Plate: Corning Costar 9018 (included with product Cat. Nos. ending in suffixes -22, -76, -86) |
| Conjugate | HRP |
| Immunoassay Kit Format | Uncoated ELISA Kit |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| Target Species | Human |
| For Use With (Application) | ELISA |
| Show More |
For Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.