missing translation for 'onlineSavingsMsg'
Learn More

CCL3/CCL4 Antibody (93342), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals FAB2701MFV450

414.80 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.


Product Code. 18559359

  • £510.00 / 0.10mL
Define Immunology Research on Your Terms With R&D Systems
Use promo code 24090
View all promotions

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

CCL3/CCL4 Monoclonal antibody specifically detects CCL3/CCL4 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CCL3/CCL4
Monoclonal
mFluor Violet 450 SE
50mM Sodium Borate
Mouse
0.1 mL
Primary
Human
IgG2b
Western Blot, Flow Cytometry, CyTOF
93342
Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready
C-C motif chemokine;C-C motif chemokine 3;G0S19-1;LD78ALPHA;MIP1A;MIP-1-alpha;SCYA3
E. coli-derived recombinant human CCL3/MIP-1 alpha , Ala27-Ala92, Accession # P10147
RUO
6348
Store at 4C in the dark.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.