Learn More
CCL3/CCL4 Antibody (93342), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB2701J
436.05 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
Product Code. 18670870
  • £547.00 / 0.10mL
Define Immunology Research on Your Terms With R&D Systems
Use promo code 24090
View all promotions
Description
CCL3/CCL4 Monoclonal antibody specifically detects CCL3/CCL4 in Human samples. It is validated for Western Blot, Flow Cytometry (Intracellular Staining), CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CCL3/CCL4
Monoclonal
Janelia Fluor 646
50mM Sodium Borate
Mouse
Protein A or G purified from ascites
RUO
6348
Store at 4°C in the dark.
IgG2b
Western Blot, Flow Cytometry (Intracellular Staining), CyTOF
93342
Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready
C-C motif chemokine;C-C motif chemokine 3;G0S19-1;LD78ALPHA;MIP1A;MIP-1-alpha;SCYA3
E. coli-derived recombinant human CCL3/MIP-1 alpha , Ala27-Ala92, Accession # P10147
0.1 mL
Primary
Human
Purified