missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCL28 Rabbit anti-Human, Clone: 12, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89585-100UL
This item is not returnable.
View return policy
Description
CCL28 Monoclonal specifically detects CCL28 in Human samples. It is validated for ELISA.
Specifications
| CCL28 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 12 | |
| ELISA 1:5000-1:10000 | |
| chemokine (C-C motif) ligand 28, member 28, MGC71902, Protein CCK1 | |
| Recombinant Human CCL28 Protein | |
| 100 μg | |
| Biologically Active Proteins | |
| 56477 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction