Learn More
PeproTech CCL28 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
Brand: PeproTech 500-P297BT-500UG
Description
AA Sequence of recombinant protein: SEAILPIASS CCTEVSHHIS RRLLERVNMC RIQRADGDCD LAAVILHVKR RRICVSPHNH TVKQWMKVQA AKKNGKGNVC HRKKHHGKRN SNRAHQGKHE TYGHKTPY. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human MEC (CCL28). Anti-Human MEC (CCL28)-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect hMEC by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human MEC (500-P297) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant hMEC. Western Blot: To detect hMEC by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hMEC is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P297BT-1MG will be provided as 2 x 500 μg
MEC is a secreted CC chemokine expressed primarily by epithelial cells of the bronchioles, salivary gland, mammary gland and colon. MEC signals through the CCR10 receptor, and chemoattracts resting CD4, CD8 T-cells and eosinophils. MEC contains six cysteines, including the four highly conserved cysteine residues present in CC chemokines.
Specifications
| CCL28 | |
| Polyclonal | |
| Biotin | |
| CCL28 | |
| CC chemokine CCL28; C-C motif chemokine 28; C-C motif chemokine ligand 28; CCK 1; CCK1; CCK1 protein; CCL 28; Ccl28; chemokine (C-C motif) ligand 28; chemokine (C-C motif) ligand 28 splice variant chi; MEC; Mucosae-associated epithelial chemokine; Protein CCK1; SCYA28; small inducible cytokine A28; small inducible cytokine subfamily A (Cys-Cys), member 28; small-inducible cytokine A28 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 56477 | |
| -20°C | |
| Lyophilized |
| ELISA, Western Blot | |
| 0.1-1.0 mg/mL | |
| PBS with no preservative | |
| Q9NRJ3 | |
| CCL28 | |
| E.coli-derived Recombinant Human MEC (CCL28). | |
| 500 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.