missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CCL28 Monoclonal Antibody (62705)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561775
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
MEC is a secreted CC chemokine expressed primarily by epithelial cells of the bronchioles, salivary gland, mammary gland and colon. MEC signals through the CCR10 receptor, and chemoattracts resting CD4, CD8 T-cells and eosinophils. MEC contains six cysteines, including the four highly conserved cysteine residues present in CC chemokines.
Specifications
| CCL28 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q9NRJ3 | |
| CCL28 | |
| E. coli-derived recombinant human CCL28, Ile23-Tyr127. | |
| 500 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Neutralization, Western Blot | |
| 62705 | |
| Unconjugated | |
| CCL28 | |
| CC chemokine CCL28; C-C motif chemokine 28; C-C motif chemokine ligand 28; CCK 1; CCK1; CCK1 protein; CCL 28; Ccl28; chemokine (C-C motif) ligand 28; chemokine (C-C motif) ligand 28 splice variant chi; MEC; Mucosae-associated epithelial chemokine; Protein CCK1; SCYA28; small inducible cytokine A28; small inducible cytokine subfamily A (Cys-Cys), member 28; small-inducible cytokine A28 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 56477 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction