missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCL28 Antibody (012), mFluor Violet 610 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89585MFV610
This item is not returnable.
View return policy
Description
CCL28 Monoclonal antibody specifically detects CCL28 in Human samples. It is validated for ELISA
Specifications
| CCL28 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 012 | |
| ELISA | |
| chemokine (C-C motif) ligand 28, member 28, MGC71902, Protein CCK1 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human CCL28 (Accession#: NP_683513.1; Ile23-Tyr127). | |
| 0.1 mL | |
| Biologically Active Proteins | |
| 56477 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction