missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCL27/CTACK Antibody (MM0152-7H44), mFluor Violet 500 SE, Novus Biologicals™
Shop All Bio Techne ProductsDescription
CCL27/CTACK Monoclonal antibody specifically detects CCL27/CTACK in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Neutralization
Specifications
Specifications
| Antigen | CCL27/CTACK |
| Applications | Neutralization |
| Classification | Monoclonal |
| Formulation | 50mM Sodium Borate |
| Immunogen | Human recombinant CCL27/CTACK |
| Purification Method | Protein G purified |
| Quantity | 0.1 mL |
| Gene ID (Entrez) | 10850 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?