missing translation for 'onlineSavingsMsg'
Learn More

CCL24/Eotaxin-2/MPIF-2 Antibody (RM0176-8M17), mFluor Violet 610 SE, Novus Biologicals™

Product Code. 18419847 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18419847 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18419847 Supplier Novus Biologicals Supplier No. NBP212109MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rat Monoclonal Antibody

CCL24/Eotaxin-2/MPIF-2 Monoclonal antibody specifically detects CCL24/Eotaxin-2/MPIF-2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Neutralization
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCL24/Eotaxin-2/MPIF-2
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Neutralization
Classification Monoclonal
Clone RM0176-8M17
Conjugate mFluor Violet 610 SE
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Block/Neutralize
Formulation 50mM Sodium Borate
Gene Alias chemokine (C-C motif) ligand 24, Ckb-6, Eosinophil chemotactic protein 2, Eotaxin-2, member 24, MPIF2Myeloid progenitor inhibitory factor 2, MPIF-2Small-inducible cytokine A24, SCYA24CK-beta-6
Host Species Rat
Immunogen Mouse recombinant CCL24/Eotaxin-2/MPIF-2
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 6369
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.