missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCL24/Eotaxin-2/MPIF-2 Antibody (MM0149-8C72), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12108MFV500
This item is not returnable.
View return policy
Description
CCL24/Eotaxin-2/MPIF-2 Monoclonal antibody specifically detects CCL24/Eotaxin-2/MPIF-2 in Human samples. It is validated for Western Blot, Neutralization
Specifications
| CCL24/Eotaxin-2/MPIF-2 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Neutralization | |
| MM0149-8C72 | |
| Western Blot, Block/Neutralize | |
| chemokine (C-C motif) ligand 24, Ckb-6, Eosinophil chemotactic protein 2, Eotaxin-2, member 24, MPIF2Myeloid progenitor inhibitory factor 2, MPIF-2Small-inducible cytokine A24, SCYA24CK-beta-6 | |
| Human recombinant CCL24/Eotaxin-2/MPIF-2 | |
| 0.1 mL | |
| Biologically Active Proteins, Cytokine Research | |
| 6369 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction