missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCL24/Eotaxin-2/MPIF-2 Antibody (MM0149-8C72), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12108AF750
This item is not returnable.
View return policy
Description
CCL24/Eotaxin-2/MPIF-2 Monoclonal specifically detects CCL24/Eotaxin-2/MPIF-2 in Human samples. It is validated for Western Blot.
Specifications
| CCL24/Eotaxin-2/MPIF-2 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| CCL24 | |
| Human recombinant CCL24/Eotaxin-2/MPIF-2 | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot | |
| MM0149-8C72 | |
| Western Blot | |
| chemokine (C-C motif) ligand 24, Ckb-6, Eosinophil chemotactic protein 2, Eotaxin-2, member 24, MPIF2Myeloid progenitor inhibitory factor 2, MPIF-2Small-inducible cytokine A24, SCYA24CK-beta-6 | |
| Mouse | |
| 0.1 mL | |
| 6369 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction