missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCL23/Ck beta 8-1/MIP3 Rabbit anti-Human, Clone: 4, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89591-50UL
This item is not returnable.
View return policy
Description
CCL23/Ck beta 8-1/MIP3 Monoclonal specifically detects CCL23/Ck beta 8-1/MIP3 in Human samples. It is validated for Sandwich ELISA Capture.
Specifications
| CCL23/Ck beta 8-1/MIP3 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 4 | |
| Sandwich ELISA Capture 1:250-1:2000 | |
| C6 beta-chemokine, chemokine (C-C motif) ligand 23, CKb8, CKB-8, CK-beta-8, hmrp-2a, Macrophage inflammatory protein 3, MIP3, MIP-3CK-BETA-8, MPIF1, MPIF-1Ckb-8-1, Myeloid progenitor inhibitory factor 1, SCYA23C-C motif chemokine 23, small inducible cytokine subfamily A (Cys-Cys), member 23, Small-inducible cytokine A23 | |
| Recombinant Human CCL23/Ck beta 8-1/MIP3 Protein | |
| 50 μg | |
| Biologically Active Proteins, Cell Cycle and Replication | |
| 6368 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction