missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CCL23/Ck beta 8-1/MIP3 Antibody (CCL23/4036) [Alexa Fluor™ 700], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27009AF700
This item is not returnable.
View return policy
Description
CCL23/Ck beta 8-1/MIP3 Monoclonal antibody specifically detects CCL23/Ck beta 8-1/MIP3 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| CCL23/Ck beta 8-1/MIP3 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| C6 beta-chemokine, chemokine (C-C motif) ligand 23, CKb8, CKB-8, CK-beta-8, hmrp-2a, Macrophage inflammatory protein 3, MIP3, MIP-3CK-BETA-8, MPIF1, MPIF-1Ckb-8-1, Myeloid progenitor inhibitory factor 1, SCYA23C-C motif chemokine 23, small inducible cytokine subfamily A (Cys-Cys), member 23, Small-inducible cytokine A23 | |
| Recombinant fragment (around aa1-120) of human CCL23/Ck beta 8-1/MIP3 protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Biologically Active Proteins, Cell Cycle and Replication | |
| 6368 | |
| Store at 4°C in the dark. | |
| IgG2c κ |
| Immunohistochemistry (Paraffin) | |
| CCL23/4036 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction