missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCL23/Ck beta 8-1/MIP3 Antibody (004), mFluor Violet 500 SE, Novus Biologicals™
Shop All Bio Techne ProductsDescription
CCL23/Ck beta 8-1/MIP3 Monoclonal antibody specifically detects CCL23/Ck beta 8-1/MIP3 in Human samples. It is validated for ELISA
Specifications
Specifications
| Antigen | CCL23/Ck beta 8-1/MIP3 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 004 |
| Conjugate | mFluor Violet 500 SE |
| Dilution | Sandwich ELISA Capture |
| Formulation | 50mM Sodium Borate |
| Gene Alias | C6 beta-chemokine, chemokine (C-C motif) ligand 23, CKb8, CKB-8, CK-beta-8, hmrp-2a, Macrophage inflammatory protein 3, MIP3, MIP-3CK-BETA-8, MPIF1, MPIF-1Ckb-8-1, Myeloid progenitor inhibitory factor 1, SCYA23C-C motif chemokine 23, small inducible cytokine subfamily A (Cys-Cys), member 23, Small-inducible cytokine A23 |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human CCL23/Ck beta 8-1/MIP3 (Accession#: AAB51134.1; Arg22-Asn120). |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?