missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCL22/MDC Antibody (RM0175-9L23), Alexa Fluor™ 350, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-12106AF350
This item is not returnable.
View return policy
Description
CCL22/MDC Monoclonal specifically detects CCL22/MDC in Mouse samples. It is validated for Western Blot.
Specifications
| CCL22/MDC | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| CCL22 | |
| Mouse recombinant CCL22/MDC | |
| 0.1 mL | |
| 6367 | |
| Store at 4°C in the dark. |
| Western Blot | |
| RM0175-9L23 | |
| Western Blot | |
| A-152E5.1, ABCD-1, CC chemokine STCP-1, C-C motif chemokine 22, chemokine (C-C motif) ligand 22, DC/B-CK, Macrophage-derived chemokine, MDCStimulated T-cell chemotactic protein 1, MGC34554, SCYA22MDC(1-69), small inducible cytokine A22, small inducible cytokine subfamily A (Cys-Cys), member 22, Small-inducible cytokine A22, STCP-1, stimulated T cell chemotactic protein 1 | |
| Rat | |
| Protein A or G purified | |
| Primary | |
| Mouse | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction