missing translation for 'onlineSavingsMsg'
Learn More

CCL22/MDC Antibody (57226) [Janelia Fluor™ 549], Novus Biologicals™

Product Code. 30736529 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30736529 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30736529 Supplier Novus Biologicals Supplier No. FAB336I

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CCL22/MDC Monoclonal antibody specifically detects CCL22/MDC in Human samples. It is validated for Western Blot,ELISA Standard (Matched Pair),Neutralization,ELISA Capture (Matched Antibody Pair),ELISA Detection (Matched Antibody Pair)
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCL22/MDC
Applications Western Blot, ELISA Standard (Matched Pair), Neutralization, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair)
Classification Monoclonal
Clone 57226
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias A-152E5.1, ABCD-1, CC chemokine STCP-1, C-C motif chemokine 22, chemokine (C-C motif) ligand 22, DC/B-CK, Macrophage-derived chemokine, MDCStimulated T-cell chemotactic protein 1, MGC34554, SCYA22MDC(1-69), small inducible cytokine A22, small inducible cytokine subfamily A (Cys-Cys), member 22, Small-inducible cytokine A22, STCP-1, stimulated T cell chemotactic protein 1
Host Species Mouse
Immunogen E. coli-derived recombinant human CCL22/MDC, Gly25-Gln93, Accession # O00626.1
Purification Method Protein A or G purified from ascites
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins
Primary or Secondary Primary
Gene ID (Entrez) 6367
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.