missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCL21/6Ckine Antibody (MM0146-2H26), mFluor Violet 610 SE, Novus Biologicals™
Shop All Bio Techne ProductsDescription
CCL21/6Ckine Monoclonal antibody specifically detects CCL21/6Ckine in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry
Specifications
Specifications
| Host Species | Mouse |
| Purification Method | Protein G purified |
| Primary or Secondary | Primary |
| Gene ID (Entrez) | 6366 |
| Isotype | IgG1 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?