missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCL21/6Ckine Antibody (MM0146-2H26), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12103MFV500
This item is not returnable.
View return policy
Description
CCL21/6Ckine Monoclonal antibody specifically detects CCL21/6Ckine in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry
Specifications
| CCL21/6Ckine | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| MM0146-2H26 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunocytochemistry | |
| 6CkineSmall-inducible cytokine A21, Beta-chemokine exodus-2, chemokine (C-C motif) ligand 21, CKb9, member 21, SCYA21MGC34555, secondary lymphoid tissue chemokine, SLCSecondary lymphoid-tissue chemokine | |
| Human recombinant CCL21/6Ckine | |
| 0.1 mL | |
| Biologically Active Proteins, Cancer, Cell Cycle and Replication, Chemokines and Cytokines | |
| 6366 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction