missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CCL20/MIP-3 alpha Antibody (GSK3050002) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28761-50ug
This item is not returnable.
View return policy
Description
CCL20/MIP-3 alpha Monoclonal antibody specifically detects CCL20/MIP-3 alpha in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| CCL20/MIP-3 alpha | |
| Monoclonal | |
| Unconjugated | |
| P78556 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG4 |
| Flow Cytometry, ELISA, Functional Assay | |
| GSK3050002 | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| beta chemokine exodus-1, Beta-chemokine exodus-1, CC chemokine LARC, C-C motif chemokine 20, chemokine (C-C motif) ligand 20, CKb4, exodus-1, LARCLiver and activation-regulated chemokine, MIP-3a, MIP-3-alpha, MIP3AMacrophage inflammatory protein 3 alpha, SCYA20Small-inducible cytokine A20, small inducible cytokine subfamily A (Cys-Cys), member 20, ST38 | |
| CCL20 | |
| 50 μg | |
| Biologically Active Proteins, Chemokines and Cytokines | |
| 6364 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction