missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CCL2/JE/MCP-1 Monoclonal antibody specifically detects CCL2/JE/MCP-1 in Human samples. It is validated for Western Blot, Neutralization
Specifications
Specifications
| Antigen | CCL2/JE/MCP-1 |
| Applications | Western Blot, Neutralization |
| Classification | Monoclonal |
| Clone | 24822 |
| Conjugate | Alexa Fluor 594 |
| Dilution | Western Blot, Neutralization |
| Formulation | 50mM Sodium Borate |
| Gene Alias | C-C motif chemokine ligand 2, CCL2, GDCF-2, HC11, HSMCR30, MCAF, Mcp1, Mcp-1, SCYA2, SMC-CF |
| Host Species | Mouse |
| Immunogen | E. coli-derived recombinant human CCL2/JE/MCP-1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?