missing translation for 'onlineSavingsMsg'
Learn More

CCL2/JE/MCP-1 Antibody (24822), Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18549918 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18549918 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18549918 Supplier Novus Biologicals Supplier No. FAB279V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CCL2/JE/MCP-1 Monoclonal antibody specifically detects CCL2/JE/MCP-1 in Human samples. It is validated for Western Blot, Neutralization
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCL2/JE/MCP-1
Applications Western Blot, Neutralization
Classification Monoclonal
Clone 24822
Conjugate Alexa Fluor 405
Dilution Western Blot, Neutralization
Formulation 50mM Sodium Borate
Gene Alias C-C motif chemokine ligand 2, CCL2, GDCF-2, HC11, HSMCR30, MCAF, Mcp1, Mcp-1, SCYA2, SMC-CF
Host Species Mouse
Immunogen E. coli-derived recombinant human CCL2/JE/MCP-1
Purification Method Protein A or G purified from ascites
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6347
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.