missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCL19/MIP-3 beta Antibody (SR1080), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22149-100ul
This item is not returnable.
View return policy
Description
CCL19/MIP-3 beta Monoclonal antibody specifically detects CCL19/MIP-3 beta in Human samples. It is validated for Western Blot, Flow Cytometry
Specifications
| CCL19/MIP-3 beta | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry | |
| SR1080 | |
| Western Blot 1:1000-1:5000, Flow Cytometry 1:100 | |
| beta chemokine exodus-3, Beta-chemokine exodus-3, CC chemokine ligand 19, C-C motif chemokine 19, chemokine (C-C motif) ligand 19, CKb11, EBI1-ligand chemokine, ELCMIP-3-beta, Epstein-Barr virus-induced molecule 1 ligand chemokine, exodus-3, Macrophage inflammatory protein 3 beta, macrophage inflammatory protein 3-beta, MGC34433, MIP-3b, MIP3BCK beta-11, SCYA19EBI1 ligand chemokine, small inducible cytokine subfamily A (Cys-Cys), member 19, Small-inducible cytokine A19 | |
| A synthesized peptide derived from human CCL19/MIP-3 beta (Uniprot #: Q99731) | |
| 100 μg | |
| Biologically Active Proteins | |
| 6363 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction