missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CCL17/TARC Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 15 publications
£170.00 - £1815.00
Specifications
| Antigen | CCL17/TARC |
|---|---|
| Clone | 110904 |
| Dilution | Neutralization 0.75-3.0 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Neutralization, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18778809
![]() |
R&D Systems
MAB529-SP |
25 μg |
£170.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18778099
![]() |
R&D Systems
MAB529-100 |
100 μg |
£447.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18763247
![]() |
R&D Systems
MAB529-500 |
500 μg |
£1815.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CCL17/TARC Monoclonal specifically detects CCL17/TARC in Mouse samples. It is validated for Neutralization, ELISA Capture (Matched Antibody Pair).Specifications
| CCL17/TARC | |
| Neutralization 0.75-3.0 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ABCD-2, CC chemokine TARC, C-C motif chemokine 17, chemokine (C-C motif) ligand 17, MGC138273, SCYA17, SCYA17MGC138271, small inducible cytokine subfamily A (Cys-Cys), member 17, Small-inducible cytokine A17, T cell-directed CC chemokine, TARC, TARCA-152E5.3, Thymus and activation-regulated chemokine | |
| CCL17 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 110904 | |
| Neutralization, ELISA | |
| Unconjugated | |
| Rat | |
| Mouse | |
| Q9WUZ6 | |
| 6361 | |
| E. coli-derived Recombinant Mouse CCL17/TARC Ala24-Pro93 Accession # Q9WUZ6 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse CCL17/TARC in ELISAs. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title