missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCL17/TARC Antibody Pair, HRP, Novus Biologicals™
Mouse Monoclonal Antibody
£292.00 - £543.00
Specifications
| Antigen | CCL17/TARC |
|---|---|
| Dilution | ELISA, Sandwich ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Conjugate | HRP |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30029170
|
Novus Biologicals
NBP3-26109-5Plates |
5 Plates |
£292.00
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30034262
|
Novus Biologicals
NBP3-26109-15Plates |
15 Plates |
£543.00
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CCL17/TARC Monoclonal antibody specifically detects CCL17/TARC in Human samples. It is validated for ELISA, ELISASpecifications
| CCL17/TARC | |
| ELISA | |
| HRP | |
| RUO | |
| Human | |
| 6361 | |
| Storage of components varies. See protocol for specific instructions. |
| ELISA, Sandwich ELISA | |
| Monoclonal | |
| Mouse | |
| Biologically Active Proteins, Chemokines and Cytokines | |
| ABCD-2, CC chemokine TARC, C-C motif chemokine 17, chemokine (C-C motif) ligand 17, MGC138273, SCYA17MGC138271, small inducible cytokine subfamily A (Cys-Cys), member 17, Small-inducible cytokine A17, T cell-directed CC chemokine, TARCA-152E5.3, Thymus and activation-regulated chemokine | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title