missing translation for 'onlineSavingsMsg'
Learn More

CCL17/TARC Antibody Pair, HRP, Novus Biologicals™

Product Code. 30029170 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
1 set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30029170 5 Plates 1 set
30034262 15 Plates 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30029170 Supplier Novus Biologicals Supplier No. NBP3261095Plates
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CCL17/TARC Monoclonal specifically detects CCL17/TARC in Human samples. It is validated for ELISA∣Sandwich ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCL17/TARC
Applications ELISA, Sandwich ELISA
Classification Monoclonal
Conjugate HRP
Dilution ELISA, Sandwich ELISA
Gene Alias ABCD-2, CC chemokine TARC, C-C motif chemokine 17, chemokine (C-C motif) ligand 17, MGC138273, SCYA17MGC138271, small inducible cytokine subfamily A (Cys-Cys), member 17, Small-inducible cytokine A17, T cell-directed CC chemokine, TARCA-152E5.3, Thymus and activation-regulated chemokine
Host Species Mouse
Quantity 5 Plates
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Chemokines and Cytokines
Primary or Secondary Primary
Gene ID (Entrez) 6361
Target Species Human
Content And Storage Storage of components varies. See protocol for specific instructions.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.