missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CCL16 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA540345
This item is not returnable.
View return policy
Description
Peptide sequence: SLLVLILIIT SASRSQPKVP EWVNTPSTCC LKYYEKVLPR RLVVGYRKAL Sequence homology: Human: 100%.
CCL16 gene is one of several cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. CCL16 displays chemotactic activity for lymphocytes and monocytes but not for neutrophils. This cytokine also shows a potent myelosuppressive activity and suppresses proliferation of myeloid progenitor cells. The expression of this gene is upregulated by IL-10.
Specifications
| CCL16 | |
| Polyclonal | |
| Unconjugated | |
| CCL16 | |
| C-C motif chemokine 16; C-C motif chemokine ligand 16; CCL16; chemokine (C-C motif) ligand 16; Chemokine CC-4; Chemokine LEC; CKb12; HCC-4; IL-10-inducible chemokine; ILINCK; LCC-1; LEC; liver CC chemokine-1; liver-expressed chemokine; LMC; Lymphocyte and monocyte chemoattractant; MGC117051; monotactin-1; MTN-1; NCC4; NCC-4; new CC chemokine 4; SCYA16; SCYL4; small inducible cytokine subfamily A (Cys-Cys), member 16; small-inducible cytokine A16 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 6360 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O15467 | |
| CCL16 | |
| Synthetic peptide directed towards the N-terminal of human CCL16 (aa 9-58). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction