Learn More
PeproTech CCL16 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
Goat Polyclonal Antibody
Brand: PeproTech 500-P125GBT-500UG
Description
AA Sequence of recombinant protein: QPKVPEWVNT PSTCCLKYYE KVLPRRLVVG YRKALNCHLP AIIFVTKRNR EVCTNPNDDW VQEYIKDPNL PLLPTRNLST VKIITAKNGQ PQLLNSQ. Preparation: Produced from sera of goats immunized with highly pure Recombinant Human LEC (CCL16). Anti-Human LEC (CCL16)-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Human LEC (CCL16) by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human LEC (CCL16) (500-P125G) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human LEC (CCL16). Western Blot: To detect hLEC by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hLEC is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P125GBT-1MG will be provided as 2 x 500 μg
CCL16 gene is one of several cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. CCL16 displays chemotactic activity for lymphocytes and monocytes but not for neutrophils. This cytokine also shows a potent myelosuppressive activity and suppresses proliferation of myeloid progenitor cells. The expression of this gene is upregulated by IL-10.
Specifications
| CCL16 | |
| Polyclonal | |
| Biotin | |
| CCL16 | |
| C-C motif chemokine 16; C-C motif chemokine ligand 16; CCL16; chemokine (C-C motif) ligand 16; Chemokine CC-4; Chemokine LEC; CKb12; HCC-4; IL-10-inducible chemokine; ILINCK; LCC-1; LEC; liver CC chemokine-1; liver-expressed chemokine; LMC; Lymphocyte and monocyte chemoattractant; MGC117051; monotactin-1; MTN-1; NCC4; NCC-4; new CC chemokine 4; SCYA16; SCYL4; small inducible cytokine subfamily A (Cys-Cys), member 16; small-inducible cytokine A16 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 6360 | |
| -20°C | |
| Lyophilized |
| ELISA, Western Blot | |
| 0.1-1.0 mg/mL | |
| PBS with no preservative | |
| O15467 | |
| CCL16 | |
| E.coli-derived Recombinant Human LEC (CCL16). | |
| 500 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.