missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CCL16 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16587801
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16587801 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16587801 Supplier Invitrogen™ Supplier No. PA590374

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: MKVSEAALSL LVLILIITSA SRSQPKVPEW VNTPSTCCLK YYEKVLPRRL VVGYRKALNC HLPAIIFVTK RNREVCTNPN DDWVQEYIKD PNLPLLPTRN LSTVKIITAK NGQPQLLNSQ; Positive Samples: Rat liver.

CCL16 gene is one of several cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. CCL16 displays chemotactic activity for lymphocytes and monocytes but not for neutrophils. This cytokine also shows a potent myelosuppressive activity and suppresses proliferation of myeloid progenitor cells. The expression of this gene is upregulated by IL-10.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCL16
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Alias C-C motif chemokine 16; C-C motif chemokine ligand 16; CCL16; chemokine (C-C motif) ligand 16; Chemokine CC-4; Chemokine LEC; CKb12; HCC-4; IL-10-inducible chemokine; ILINCK; LCC-1; LEC; liver CC chemokine-1; liver-expressed chemokine; LMC; Lymphocyte and monocyte chemoattractant; MGC117051; monotactin-1; MTN-1; NCC4; NCC-4; new CC chemokine 4; SCYA16; SCYL4; small inducible cytokine subfamily A (Cys-Cys), member 16; small-inducible cytokine A16
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-120 of human CCL16 (NP_0045811).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.