missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCL16/HCC-4/LEC Antibody (013), DyLight 405 , Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89529V
This item is not returnable.
View return policy
Description
CCL16/HCC-4/LEC Monoclonal antibody specifically detects CCL16/HCC-4/LEC in Human samples. It is validated for ELISA
Specifications
| CCL16/HCC-4/LEC | |
| Monoclonal | |
| DyLight 405 | |
| C-C motif chemokine 16, chemokine (C-C motif) ligand 16, Chemokine CC-4, Chemokine LEC, CKb12, HCC-4Liver-expressed chemokine, LCC-1Lymphocyte and monocyte chemoattractant, LEC, liver CC chemokine-1, LMCSmall-inducible cytokine A16, monotactin-1, MTN-1, NCC4IL-10-inducible chemokine, NCC-4ILINCK, new CC chemokine 4, SCYA16MGC117051, SCYL4, small inducible cytokine subfamily A (Cys-Cys), member 16 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human CCL16/HCC-4/LEC (NP_004581.1; Gln24-Gln120). | |
| 0.1 mL | |
| Biologically Active Proteins, Chemokines and Cytokines | |
| 6360 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 013 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction