missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCL15/MIP-1 delta Rabbit anti-Human, Clone: 124, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89704-100UL
This item is not returnable.
View return policy
Description
CCL15/MIP-1 delta Monoclonal specifically detects CCL15/MIP-1 delta in Human samples. It is validated for Sandwich ELISA Capture.
Specifications
| CCL15/MIP-1 delta | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 124 | |
| Sandwich ELISA Capture 1:250-1:2000 | |
| chemokine (C-C motif) ligand 15, Chemokine CC-2, HCC-2Lkn-1, leukotactin 1, Leukotactin-1, LKN1, LKN-1, Macrophage inflammatory protein 5, MIP-1 delta, MIP5, MIP-5MIP-1d, Mrp-2b, NCC-3MIP-1D, NCC3MRP-2B, new CC chemokine 3, SCYA15member 15, Small-inducible cytokine A15 | |
| Recombinant Human CCL15/MIP-1 delta Protein | |
| 100 μg | |
| Biologically Active Proteins, Chemokines and Cytokines, Protein Kinase | |
| 6359 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction