missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CCL14 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA556421
This item is not returnable.
View return policy
Description
Immunogen sequence: TKTESSSRGP YHPSECCFTY TTYKIPRQRI MDYYETNSQC SKPGIVFITK RGHSVCTNPS DKWVQDYIKD MKE Highest antigen sequence identity to the following orthologs: Mouse - 47%, Rat - 45%.
This gene, CCL14, is one of several CC cytokine genes clustered on 17q11.2. The CC cytokines are secreted proteins characterized by two adjacent cysteines. The cytokine encoded by this gene induces changes in intracellular calcium concentration and enzyme release in monocytes. This gene expresses both monocistronic and bicistronic transcripts. Bicistronic transcripts include the upstream cytokine gene CCL15. In addition, CCL14 undergoes alternative splicing in the bicistronic transcripts; it is unknown if its monocistronic transcript is also subject to alternative splicing.
Specifications
| CCL14 | |
| Polyclonal | |
| Unconjugated | |
| CCL14 | |
| C-C motif chemokine 14; C-C motif chemokine ligand 14; CC-1; CC-3; CCL14; chemokine (C-C motif) ligand 14; chemokine CC-1/CC-3; chemokine CC-3; CKB1; H HCC-1(1-74); HCC-1; HCC-1(1-74); HCC-1(3-74); HCC-1(4-74); HCC-1(9-74); HCC-1/HCC-3; HCC-3; hemofiltrate CC chemokine 1; MCIF; NCC2; NCC-2; new CC chemokine 2; SCYA14; SCYL2; small inducible cytokine subfamily A (Cys-Cys), member 14; small-inducible cytokine A14; SY14 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6358 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q16627 | |
| CCL14 | |
| Recombinant protein corresponding to Human CCL14. Recombinant protein control fragment (Product #RP-95149). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction