missing translation for 'onlineSavingsMsg'
Learn More

CCL14/HCC-1/HCC-3 Antibody (256409R) [Janelia Fluor™ 646], Novus Biologicals™

Product Code. 30734627 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30734627 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30734627 Supplier Novus Biologicals Supplier No. FAB3241RJ

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

CCL14/HCC-1/HCC-3 Monoclonal antibody specifically detects CCL14/HCC-1/HCC-3 in Human samples. It is validated for Western Blot,ELISA Standard (Matched Pair),Neutralization,ELISA Capture (Matched Antibody Pair),ELISA Detection (Matched Antibody Pair)
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCL14/HCC-1/HCC-3
Applications Western Blot, ELISA Standard (Matched Pair), Neutralization, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair)
Classification Monoclonal
Clone 256409R
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias C-C motif chemokine 14, chemokine (C-C motif) ligand 14, Chemokine CC-1/CC-3, chemokine CC-3, CKB1, FLJ16015, HCC-1, HCC-1(1-74), HCC-1/HCC-3, HCC-3, hemofiltrate CC chemokine 1, MCIF, member 14, NCC2CC-1, NCC-2CKb1, new CC chemokine 2, SCYA14CC-3, Small-inducible cytokine A14
Host Species Rat
Immunogen E. coli-derived recombinant human CCL14/HCC-1/HCC-3, Thr20-Asn109, Accession # NP_116738
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cell Cycle and Replication, Chemokines and Cytokines
Primary or Secondary Primary
Gene ID (Entrez) 6358
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.