missing translation for 'onlineSavingsMsg'
Learn More

CCL12/MCP-5 Antibody (RM0169-8J25), Janelia Fluor™ 549, Novus Biologicals™

Codice prodotto. 18422516 Sfoglia Tutto Bio Techne Prodotti
Cambia vista
Click to view available options
Quantity:
0.1 mL
Dimensione della confezione:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantity unitSize
18422516 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 18422516 Fornitore Novus Biologicals N. del fornitore NBP212089JF549

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Rat Monoclonal Antibody

CCL12/MCP-5 Monoclonal antibody specifically detects CCL12/MCP-5 in Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifica

Antigen CCL12/MCP-5
Applications Western Blot
Classification Monoclonal
Clone RM0169-8J25
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias C-C motif chemokine 12, chemokine (C-C motif) ligand 12, MCP-5, Scya12
Host Species Rat
Immunogen Mouse recombinant CCL12/MCP-5
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins
Primary or Secondary Primary
Gene ID (Entrez) 20293
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2
Vedi altri risultati Mostra meno risultati
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.