missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCL1/I-309/TCA-3 Antibody (RM0167-6H50), Alexa Fluor™ 647, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-11876AF647
This item is not returnable.
View return policy
Description
CCL1/I-309/TCA-3 Monoclonal specifically detects CCL1/I-309/TCA-3 in Mouse samples. It is validated for Western Blot.
Specifications
| CCL1/I-309/TCA-3 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CCL1 | |
| Mouse recombinant CCL1/I-309/TCA-3 | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot | |
| RM0167-6H50 | |
| Western Blot | |
| C-C motif chemokine 1, chemokine (C-C motif) ligand 1, I-309, inflammatory cytokine I-309, P500, SCYA1Small-inducible cytokine A1, SISe, small inducible cytokine A1 (I-309, homologous to mouse Tca-3), T lymphocyte-secreted protein I-309, TCA3 | |
| Rat | |
| Protein A or G purified | |
| Chemokines and Cytokines | |
| 6346 | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction