missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CCKBR Polyclonal Antibody

Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Orangutan, Gibbon (100%) Monkey (83%).
CCKBR is a G-protein coupled receptor for gastrin and cholecystokinin (CCK), regulatory peptides of the brain and gastrointestinal tract. This protein is a type B gastrin receptor, which has a high affinity for both sulfated and nonsulfated CCK analogs and is found principally in the central nervous system and the gastrointestinal tract. A misspliced transcript variant including an intron has been observed in cells from colorectal and pancreatic tumors. CCKBR has been reported in brain, colon, pancreas, stomach, thyroid, and various cancers.
Specifications
Specifications
| Antigen | CCKBR |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide |
| Gene | CCKBR |
| Gene Accession No. | P32239 |
| Gene Alias | CCK(B) receptor; CCK2 receptor; Cck2r; CCK2-R; CCKB; CCK-B; Cckb receptor; CCK-B receptor; Cckbr; CCK-BR; CCKB-R; CCKR-2; CCKRB; cholecystokinin B receptor; cholecystokinin-2 receptor; Cholecystokinin-B; Cholrec; GASR; gastrin receptor; gastrin/cholecystokinin type B receptor; HGNC:1571 |
| Gene Symbols | CCKBR |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?