missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCK Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13074-2-AP-20UL
This item is not returnable.
View return policy
Description
Cholecystokinin (CCK) is a neuropeptide hormone and neurotransmitter widely distributed throughout the gastrointestinal (GI) tract and the central nervous system (CNS). Antibodies that react specifically with CCK are useful for the study of the mode of action, differential tissue expression and intracellular and subcellular localization of CCK in the CNS and periphery as well as in neuroendocrine cells of the digestive system.Specifications
| CCK | |
| Polyclonal | |
| Unconjugated | |
| CCK | |
| CCK, CCK12 Cholecystokinin 8, CCK18 Cholecystokinin 12, CCK25 Cholecystokinin 18, CCK33 Cholecystokinin 25, CCK39 Cholecystokinin 33, CCK5, CCK7 Cholecystokinin 5, CCK8 Cholecystokinin 7, cholecystokinin | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 885 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P06307 | |
| CCK | |
| CCK Fusion Protein Ag3725 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction