missing translation for 'onlineSavingsMsg'
Learn More

Motic™ CCIS™ Plan Achromatic Objective UC

Color corrected infinity optical system/plan achromat objective for crisp, flat, high-contrast images.

Brand:  Motic™ 1101001705241

Additional Details : Weight : 0.20000kg

 View more versions of this product

Product Code. 15960571

  • £144.00 / Each
Estimated Shipment: 15-07-2024
to see stock.
Add to basket

Description

Description

  • Color corrected infinity system.
    • Brilliant, flawless image quality.
    • Image aberrations are corrected right to the edge of the field of view.
  • Plan achromat lenses.
    • Provide field flatness.
  • Suitable for the following:
    • Panthera U Binocular
    • Panthera U Trinocular
    • Panthera KU Binocular
    • Panthera KU Trinocular
    • Panthera C Binocular
    • Panthera C Trinocular
    • Panthera CC Binocular
    • Panthera CC Trinocular
    • Panthera DL
    • Panthera L
Specifications

Specifications

Objective type: Plan achromatic
Objective NA: 0.80, WD: 0.35 mm, DIN: 45 mm
Objective tube length: Infinity CCIS™
Objective mounting thread: W 4/5 in x 1/36 in. (RMS standard)
Panthera series microscopes
60X
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers