missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCG-203971, MedChemExpress
Click to view available options
Quantity:
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
Description
CCG-203971 is a second-generation Rho/MRTF/SRF pathway inhibitor. CCG-203971 potently targets RhoA/C-activated SRE-luciferase (IC50 =6.4 μM). CCG-203971 inhibits PC-3 cell migration with an IC50 of 4.2 μM. Potential anti-metastasis Agent.
Specifications
Specifications
| Chemical Name or Material | CCG-203971 |
| CAS | 1443437-74-8 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C23H21ClN2O3 |
| Quantity | 100 mg |
| Solubility Information | DMSO : 250 mg/mL (611.43 mM; Need ultrasonic) |
| SMILES | O=C(C1CN(C(C2=CC=CC(C3=CC=CO3)=C2)=O)CCC1)NC4=CC=C(Cl)C=C4 |
| Molecular Weight (g/mol) | 408.88 |
| Formula Weight | 408.88 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction