missing translation for 'onlineSavingsMsg'
Learn More

CCG 203769, MedChemExpress

Product Code. 16464096
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 16464096

Brand: Medchem Express HYU00431

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CCG 203769 is a selective G protein signaling (RGS4) inhibitor, which blocks the RGS4-Gαo protein-protein interaction in vitro with an IC50 of 17 nM.

CCG 203769 is a selective G protein signaling (RGS4) inhibitor, which blocks the RGS4-Gαo protein-protein interaction in vitro with an IC50 of 17 nM.

Specifications

Chemical Name or Material CCG 203769
CAS 410074-60-1
Color Colorless
Purity Grade Notes Research
Molecular Formula C8H14N2O2S
Quantity 25 mg
Solubility Information DMSO : 62.5 mg/mL (308.99 mM; Need ultrasonic)
SMILES O=C(N1CCCC)N(CC)SC1=O
Molecular Weight (g/mol) 202.27
Formula Weight 202.27
Percent Purity 99.62%
Grade Research
Physical Form Liquid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.