missing translation for 'onlineSavingsMsg'
Learn More

CCF4-AM

Product Code. 10226212
Click to view available options
Quantity:
5mg
20mg
Unit Size:
20mg
5mg
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10226212

Brand: Invitrogen K1029

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

CCF4-AM

CCF4-AM, our LiveBLAzer™ substrate, is a lipophilic, esterified form of the CCF4 substrate which allows it to readily enter cells. Upon entry, cleavage by endogenous cytoplasmic esterases rapidly converts CCF4-AM into its negatively charged form, CCF4, which is retained in the cytosol. CCF4 is a Fluorescence Resonance Energy Transfer (FRET) substrate which consists of a cephalosporin core linking 7-hydroxycoumarin to fluorescein. This product is to be used as a FRET-based substrate for β-lactamase as a sensitive reporter of mammalian gene expression in the GeneBLAzer™ technology.
TRUSTED_SUSTAINABILITY

Specifications

Cell Permeability Cell-Permeant
Content And Storage Store in freezer (-5 to -30°C).
Detection Method Fluorescence
Form Solid
Product Line LiveBLAzer
Quantity 20mg
Substrate CCF4-AM
Substrate Properties Chemical Substrate
Substrate Type Beta-Lactamase Substrate
For Use With (Equipment) Flow Cytometer, Microplate Reader
Shipping Condition Room Temperature
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.