missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCF2-AM
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Description
CCF2-AM, our LiveBLAzer™ substrate, is a lipophilic, esterified form of the CCF2 substrate which allows it to readily enter cells. Upon entry, cleavage by endogenous cytoplasmic esterases rapidly converts CCF2-AM into its negatively charged form, CCF2 or CCF4, which is retained in the cytosol. CCF2 is a Fluorescence Resonance Energy Transfer (FRET) substrate which consists of a cephalosporin core linking 7-hydroxycoumarin to fluorescein. This product is to be used as a FRET-based substrate for β-lactamase as a sensitive reporter of mammalian gene expression in the GeneBLAzer™ technology.
Specifications
Specifications
| Cell Permeability | Cell-Permeant |
| Content And Storage | Store in freezer (-5 to -30°C). |
| Detection Method | Fluorescence |
| Product Line | GeneBLAzer |
| Quantity | 5 mg |
| Substrate | CCF2-AM |
| Substrate Properties | Chemical Substrate |
| Substrate Type | Beta-Lactamase Substrate |
| For Use With (Equipment) | Flow Cytometer, Microplate Reader |
| Shipping Condition | Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction