missing translation for 'onlineSavingsMsg'
Learn More

CCDC99 Antibody (2F4), Novus Biologicals™

Product Code. 18329109 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18329109 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18329109 Supplier Novus Biologicals Supplier No. H00054908M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CCDC99 Monoclonal antibody specifically detects CCDC99 in Human, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC99
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 2F4
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH12568
Gene Alias Arsenite-related gene 1 protein, coiled-coil domain containing 99, Coiled-coil domain-containing protein 99, FLJ20364, FLJ40690, hSpindly, protein Spindly, Rhabdomyosarcoma antigen MU-RMS-40.4A, rrhabdomyosarcoma antigen protein MU-RMS-40.4A, Spindly
Host Species Mouse
Immunogen CCDC99 (AAH12568, 1 a.a. ~ 605 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MEADIITNLRCRLKEAEEERLKAAQYGLQLVESQNELQNQLDKCRNEMMTMTESYEQEKYTLQREVELKSRMLESLSCECEAIKQQQKMHLEKLEEQLSRSHGQEVNELKTKIEKLKVELDEARLSEKQLKHQVDHQKELLSCKSEELRVMSERVQESMSSEMLALQIELTEMESMKTTLKEEVNELQYRQEQLELLITNLMRQVDRLKEEKEEREKEAVSYYNALEKARVANQDLQVQLDQALQQALDPNSKGNSLFAEVEDRRAAMERQLISMKVKYQSLKKQNVFNREQMQRMKLQIATLLQMKGSQTEFEQQERLLAMLEQKNGEIKHLLGEIRNLEKFKNLYDSMESKPSVDSGTLEDNTYYTDLLQMKLDNLNKEIESTKGELSIQRMKALFESQRALDIERKLFANERCLQLSESENMKLRAKLDELKLKYEPEETVEVPVLKKRREVLPVDITTAKDACVNNSALGGEVYRLPPQKEETQSCPNSLEDNNLQLEKSVSIHTPVVSLSPHKNLPVDMQLKKEKKCVKLIGVPADAEALSERSGNTPNSPRLAAESKLQTEVKEGKETSSKLEKETCKKSHPILYVSSKSTPETQCPQQ
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54908
Target Species Human, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.