missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CCDC97 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA532015
This item is not returnable.
View return policy
Description
Recommended positive controls: Jurkat. Predicted reactivity: Mouse (93%), Rat (93%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CCDC97 is a coiled-coil domain containing protein.
Specifications
| CCDC97 | |
| Polyclonal | |
| Unconjugated | |
| CCDC97 | |
| CCDC97; coiled-coil domain containing 97; coiled-coil domain-containing protein 97 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 90324 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.53 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q96F63 | |
| CCDC97 | |
| Recombinant protein encompassing a sequence within the center region of human CCDC97. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido