missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC92 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92829-0.1ml
This item is not returnable.
View return policy
Description
CCDC92 Polyclonal antibody specifically detects CCDC92 in Human samples. It is validated for Western Blot
Specifications
| CCDC92 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| coiled-coil domain containing 92, coiled-coil domain-containing protein 92, FLJ22471, limkain beta 2, Limkain beta-2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-331 of human CCDC92 (NP_001291889.1). MTSPHFSSYDEGPLDVSMAATNLENQLHSAQKNLLFLQREHASTLKGLHSEIRRLQQHCTDLTYELTVKSSEQTGDGTSKSSELKKRCEELEAQLKVKENENAELLKELEQKNAMITVLENTIKEREKKYLEELKAKSHKLTLLSSELEQRASTIAYLTSQLHAAKKKLMSSSGTSDASPSGSPVLASYKPAPPKDKLPETPRRRMKKSLSAPLHPEFEEVYRFGAESRKLLLREPVDAMPDPTPFLLARESAEVHLIKERPLVIPPIASDRSGEQHSPAREKPHKAHVGVAHRIHHATPPQAQPEVKTLAVDQVNGGKVVRKHSGTDRTV | |
| 0.1 mL | |
| Cell Biology, Cell Cycle and Replication | |
| 80212 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction