missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC91 Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98667JF646
This item is not returnable.
View return policy
Description
CCDC91 Polyclonal antibody specifically detects CCDC91 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| CCDC91 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 55297 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Janelia Fluor 646 | |
| coiled-coil domain containing 91, coiled-coil domain-containing protein 91, DKFZp779L1558, FLJ11088, GGA binding partner, GGA-binding partner, GGABP, p56, p56 accessory protein | |
| Produced in rabbits immunized with E. coli-derived Human CCDC91 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction