missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC83 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20902-1-AP-20UL
This item is not returnable.
View return policy
Specifications
| CCDC83 | |
| Polyclonal | |
| Unconjugated | |
| CCDC83 | |
| CCDC83, HSD9 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 220047 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.47 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8IWF9 | |
| CCDC83 | |
| CCDC83 Fusion Protein Ag15023 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction